Order Entry
Austria
ContactUsLinkComponent
Hydrazine Sulphate Solution
Hydrazine Sulphate Solution
Catalog # CMPLOP002
New Product
CAS Number:  
Hydrazine Sulphate Solution
Catalog # CMPLOP002
New Product
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Hydrazine sulphate solution
  • Shelf life:
    6 Months
  • Storage temperature:
    Store under normal laboratory conditions, at temperatures between 15...25 °C

Specifications

About this item

Certifications: ISO 9001 certified.