Human Poliovirus Receptor (PVR) (NM_006505) ORF Clone Lentiviral Particle
ORIGRC202254L3V
: OriGene
New Product
:
- Pk:200 µl
- Species:Human
- Protein synonyms:TAGE4|PVS|CD155|NECL5|Necl-5|HVED
- UniProtKB:P15151
- Protein/peptide name:Poliovirus Receptor
- Molecular weight:45,3 kDa