Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR® Fixing Solution

VWR® Fixing Solution

Fixing solution, Plastic bottle, 500 mL
Catalog # 87969.260
Supplier:   VWR Chemicals
VWR® Fixing Solution
Catalog # 87969.260
Supplier:  VWR Chemicals
CAS Number:  
VWR® Fixing Solution
VWR® Fixing Solution

Fixing solution, Plastic bottle, 500 mL

CAS Number:  
Price
158,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Fixing solution with 0,02% formaldehyde and 0,01% citric acid in aqueous solution.

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Fixing solution

Specifications