Zinc chloride 20% in formic acid 68% Reag. Ph. Eur. 1096601
Catalog # 87965.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
CAS Number:7646-85-7
:
Formula:ZnCl₂
MDL Number:MFCD00011295
:136.3 g/mol
Boiling Pt:732
Melting Pt:283
EINECS:231-592-0
UN:1760
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
ADR:8 II
Merck Index:13,10185