Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium hydroxide
Potassium hydroxide 3% in ethanol 95% Reag. Ph. Eur. 1070303
Potassium hydroxide 3% in ethanol 95% Reag. Ph. Eur. 1070303
Potassium hydroxide 3% in ethanol 95% Reag. Ph. Eur. 1070303
87906.180
:
VWR Chemicals
:
Potassium hydroxide 3% in ethanol 95% Reag. Ph. Eur. 1070303
87906.180
:
VWR Chemicals
:
Pk:
100 mL
Seq:
2440
:
:
56.11 g/mol
:
1310-58-3
:
KOH
:
MFCD00003553