Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Mercury (II) nitrate
Mercury(II) nitrate ≥70% in aqueous solution Reag. Ph. Eur. 1052801
Mercury(II) nitrate 70% ≥70% in aqueous solution Reag. Ph. Eur. 1052801
Mercury(II) nitrate 70% ≥70% in aqueous solution Reag. Ph. Eur. 1052801
87867.180
:
VWR Chemicals
:
Mercury(II) nitrate 70% ≥70% in aqueous solution Reag. Ph. Eur. 1052801
87867.180
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
100 mL
Seq:
2440
:
:
324.60 g/mol
:
233-152-3
:
3287
:
6.1,II
:
GHS06
:
GHS08
:
GHS09
:
10045-94-0
:
HgN₂O₆
:
MFCD00011038