Order Entry
Austria
ContactUsLinkComponent
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l in aqueous solution Reag. Ph. Eur. 1038100 redox indicator
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l in aqueous solution Reag. Ph. Eur. 1038100 redox indicator
Catalog # 87702.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l in aqueous solution Reag. Ph. Eur. 1038100 redox indicator
Catalog # 87702.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :692.54 g/mol
Density:1.01 g/cm³ (20 °C)
EINECS:238-676-6
CAS Number:14634-91-4
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428