Order Entry
Austria
ContactUsLinkComponent
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 1078302
Corrosive
Environment
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 1078302
Catalog # 87925.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Environment
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 1078302
Catalog # 87925.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:3264
ADR:8,III
 :GHS05
 :GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414