Order Entry
Austria
Orders LinkContactUsLinkComponent
Celite® 545, filter aid

Celite® 545, filter aid

Celite® 545, filter aid
Certificates
Celite® 545, filter aid

3 Options Available Below

Product Details & Documents
Synonyms:  DE, Diatomaceous earth, flux-calcined
  • CAS Number:68855-54-9
  • Formula:SiO₂
  • MDL Number:MFCD00132803
  • Molecular Weight:60.08 g/mol
  • Storage Temperature:Ambient
  • EINECS:272-489-0
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Particle size:
    ≤25 % (Retained on 150 mesh)
  • Wet density:
    255 to 370 g/l
  • pH:
    9.0 to 11.0 (10 % slurry)
  • Permeability:
    2.6 to 6.5 Darcy
  • Calcium oxide (CaO):
    0.3 % (Typical)
  • Magnesium oxide (MgO):
    0.3 % (Typical)
  • Silicon dioxide (SiO2):
    91.5 % (Typical)
  • Aluminium oxide (Al2O3):
    1.0 % (Typical)
  • Iron(III)oxide (Fe2O3):
    1.5 % (Typical)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 47,40 €
    47,40 €
    1 * 500 g
       47,40 €

  • Plastic drum
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 158,00 €
    158,00 €
    1 * 2,5 kg
       158,00 €

  • Fibre drum
    10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 268,00 €
    268,00 €
    1 * 10 kg
       268,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.