Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Conductivity Standards
Conductivity standard 1413 µS/cm for conductivity measurement
Conductivity standard 1413 µS/cm for conductivity measurement
Conductivity standard 1413 µS/cm for conductivity measurement
Catalog # KNICCS-C1413K/500
Supplier:
KNICK
CAS Number:
Expand image
Conductivity standard 1413 µS/cm for conductivity measurement
Catalog # KNICCS-C1413K/500
Supplier:
KNICK
CAS Number:
Specifications
Pk:
500 mL
Reference temperature:
25 °C
Description:
KCl solution
Conductivity:
1,413 µS/cm
Specifications
About this item
Product Details
Product Details