ContactUsLinkComponent
Conductivity standard 12880 µS/cm (25 °C; 0,1 mol/L; KCl) for conductivity measurement
  SCOI285126528
 :  
Conductivity standard 12880 µS/cm (25 °C; 0,1 mol/L; KCl) for conductivity measurement
  SCOI285126528
 :  
  • Pack type:
    Glass ampoule
  • Pk:
    18 Ampoul
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard 12 880 µS/cm (0.1 mol/l; KCl)