ContactUsLinkComponent
Conductivity standard 1000 µS/cm (25 °C; 0,007 mol/L; KCl) for conductivity measurement
Conductivity standard 1000 µS/cm (25 °C; 0,007 mol/L; KCl) for conductivity measurement
Catalog # YSIA060907
Supplier:  YSI ENVIRONMENTAL
CAS Number:  
Conductivity standard 1000 µS/cm (25 °C; 0,007 mol/L; KCl) for conductivity measurement
Catalog # YSIA060907
Supplier:  YSI ENVIRONMENTAL
CAS Number:  
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    475 mL
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard 1000 µS/cm (0,007 mol/l; KCl)