ContactUsLinkComponent
Conductivity standard 10000 µS/cm (25 °C; 0,0765 mol/L; KCl) for conductivity measurement
Conductivity standard 10000 µS/cm (25 °C; 0,0765 mol/L; KCl) for conductivity measurement
  YSIA065272
 :  
Conductivity standard 10000 µS/cm (25 °C; 0,0765 mol/L; KCl) for conductivity measurement
  YSIA065272
 :  
  • Pk:
    1
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard 10 000 µS/cm (0,0765 mol/l; KCl)