Order Entry
Austria
ContactUsLinkComponent
Iron(III) chloride 13 g/l in aqueous solution Reag. Ph. Eur. 1037802
Irritant
Iron(III) chloride 13 g/l in aqueous solution Reag. Ph. Eur. 1037802
Catalog # 87834.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Iron(III) chloride 13 g/l in aqueous solution Reag. Ph. Eur. 1037802
Catalog # 87834.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :162.21 g/mol
EINECS:231-729-4
UN:2582
ADR:8,III
 :GHS07
CAS Number:7705-08-0
Formula:FeCl₃
MDL Number:MFCD00011005