

Sudan I, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:842-07-9
:
Formula:C₁₆H₁₂N₂O
MDL Number:MFCD00003911
:248.28 g/mol
Boiling Pt:444 °C (1013 hPa)
Melting Pt:131…133 °C
Density:1,119 g/cm³ (151 °C)
EINECS:212-668-2
:GHS07
:GHS08
Specification Test Results
- Appearance:Orange to red to brown Powder
- Infrared spectrum:Conforms
- UV Lambda max:at approximately 418 and 476 nm (in ethanol)
- Specific extinction E (1%/1cm):≥500 (at 476 nm, in ethanol)
Specification Test Results
Frequently Bought Together