Sudan Red G
Catalog # EHERC16986127
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:278.31 g/mol
Melting Pt:225 °C
EINECS:214-968-9
CAS Number:1229-55-6
Formula:C₁₇H₁₄N₂O₂
MDL Number:MFCD00046377