Sudan red G, Supelco®
Catalog # 91282-25MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:278.31 g/mol
Melting Pt:225 °C
:M-Clarity™ quality level = MQ100
EINECS:214-968-9
CAS Number:1229-55-6
Formula:C₁₇H₁₄N₂O₂
MDL Number:MFCD00046377