Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Picric acid
Picric acid 13 g/l saturated solution in water
Picric acid 13 g/l saturated solution in water
Picric acid 13 g/l saturated solution in water
CTEH640760
:
CLIN TECH
:
Picric acid 13 g/l saturated solution in water
CTEH640760
:
CLIN TECH
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
229.11 g/mol
:
13,07492
:
201-865-9
:
88-89-1
:
C₆H₃N₃O₇
:
MFCD00007102