4-Acetylbenzonitrile 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1443-80-7
:
Formula:C₉H₇NO
MDL Number:MFCD00001825
:145.16
Boiling Pt:110 - 112 °C (1 -rr)
Melting Pt:55…58 °C
Storage Temperature:Refrigerator
EINECS:215-885-0
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Light yellow to beige Crystalline powder or mass and/or chunks
- Infrared spectrum:Conforms
- Melting point:55 to 58 °C
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together