Order Entry
Austria
ContactUsLinkComponent
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, BioChemica®
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, BioChemica®
Supplier:  PanReac AppliChem
Certificates
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, BioChemica®
Supplier:  PanReac AppliChem
Certificates

Specification Test Results

  • Assay (HPLC):
    min. 99 %
  • α20°C/D; 1 %, H2O:
    -28.5° - -34.5°
  • pH (5 %; H2O):
    5.0 - 7.0
  • Dioxane:
    max. 0.0001 %
  • Water (K.F.):
    max. 1.0 %

Specification Test Results

Related Information

Frequently Bought Together