IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99% (by HPLC), white powder for genomics
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside
CAS Number:367-93-1
:
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273
:238.31 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:110 °C (lit.)
Storage Temperature:Refrigerator
EINECS:206-703-0
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- Purity:>99%
- Optical Rotation (c=1,
water):-30° to -33° - Dioxane Content:<0.0001%
- Water Content (KF):<1.0%
- RNAse,DNAse, Protease:None Detected
Specification Test Results
Frequently Bought Together