Order Entry
Austria
ContactUsLinkComponent
 
IPTG (isopropyl-β-D-thiogalactopyranoside)
Catalog # MOLEM76948036
Supplier:  Molekula
CAS Number:  
IPTG (isopropyl-β-D-thiogalactopyranoside)
Catalog # MOLEM76948036
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside
 :
 :238.31 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:109.5…110.5 °C
Storage Temperature:Freezer
EINECS:206-703-0
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273