Orders LinkContactUsLinkComponent
Temperature Data Logger for Process Validation, EBI 12-T240
Temperature Data Logger for Process Validation, EBI 12-T240
Supplier:   EBRO
Temperature Data Logger for Process Validation, EBI 12-T240
Supplier:  EBRO
Certificates
Temperature Data Logger for Process Validation, EBI 12-T240
2 Options Available Below
Product Details & Documents
The EBI 12–T240 is a precise temperature data logger for process validation in laboratories and the pharmaceutical industry. With a temperature range of –200 to +200 °C and an accuracy of up to 0,05 °C, it enables precise temperature measurements in critical applications such as sterilisation, disinfection and ovens.
  • Suitable for steam sterilisers, washer–disinfectors
  • Temperature range –200 to +200 °C
  • High accuracy up to 0,05 °C
  • One bendable metal probe (Ø 1,50 mm)
The EBI 12–T240 temperature data logger from ebro fulfils the stringent requirements of the pharmaceutical industry and laboratory applications. It is ideal for process validation in steam sterilisers, washer-disinfectors and other critical processes where precise temperature control is required. With a wide temperature range from –200 to +200 °C and an accuracy of up to 0,05 °C, it ensures reliable results. Thanks to real-time data transmission and flexible measurement interval selection, it is perfect for seamless documentation in validated processes. Its robust design ensures a long service life and high resistance in demanding environments.
Calibration points: –20 °C, 0 °C, +60 °C and +134 °C
Operating temperature: –90 to +150 °C
Dimensions: Ø 48×24 mm H, probe length 250 or 500 mm
Certifications: Certified according to ISO 17665, ISO 15883, (EC) 1935/2004​.
Delivery information: Includes 1×EBI 12-T240, 1×factory calibration certificate, manual.
Specifications for Product Family
Specifications
  • Catalog No:
  • EBRO1340-6611
  • EBRO1340-6612
  • Model:
  • EBI 12-T240
  • EBI 12-T241
  • Sensor:
  • PT1000, class A
    Radial, 1 external temperature probe, bendeble, Ø 1,5 mm, 250 mm
  • PT1000, class A
    Radial, 1 external temperature probe, bendeble, Ø 1,5 mm, 500 mm
  • Protection class:
  • IP 68
  • IP 68
  • Recording intervals:
  • 250 ms to 24 hrs.
  • 250 ms to 24 hrs.
  • Weight:
  • 90 g
  • 90 g
  • Data memory - No. of readings:
  • 100,320
  • 100,320
  • Batteries:
  • Lithium, 2× BR1225A, 3 V user–replaceable
  • Lithium, 2× BR1225A, 3 V user–replaceable
Specifications
Product Family Options
Data loggers
Product Information
  • Range
    Accuracy
    Resolution
    Availability
    Price
  • EBRO1340-6611
    EBI 12-T240, high temperature data logger with two bendable probes, 250 mm length
    EBI 12-T240, high temperature data logger with two bendable probes, 250 mm length
    –200…+200 °C
    ±1,5 °C (–200…–90 °C)
    ±0,5 °C (–90…–40 °C)
    ±0,2 °C (–40…0 °C)
    ±0,1 °C (0…+120 °C)
    ±0,05 °C (+120…+140 °C)
    ±0,1 °C (+140…+150 °C)
    ±0,5 °C (+150…+200 °C)
    0,01 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 090,00 €
    3 090,00 €
    1 * 1 ST
       3 090,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    EBRO1340-6611
    Model
    EBI 12-T240
    Sensor
    PT1000, class A
    Radial, 1 external temperature probe, bendeble, Ø 1,5 mm, 250 mm
    Protection class
    IP 68
    Recording intervals
    250 ms to 24 hrs.
    Weight
    90 g
    Data memory - No. of readings
    100,320
    Batteries
    Lithium, 2× BR1225A, 3 V user–replaceable
  • EBRO1340-6612
    EBI 12-T241, high temperature data logger with two bendable probe, 500 mm length
    EBI 12-T241, high temperature data logger with two bendable probe, 500 mm length
    –200…+200 °C
    ±1,5 °C (–200…–90 °C)
    ±0,5 °C (–90…–40 °C)
    ±0,2 °C (–40…0 °C)
    ±0,1 °C (0…+120 °C)
    ±0,05 °C (+120…+140 °C)
    ±0,1 °C (+140…+150 °C)
    ±0,5 °C (+150…+200 °C)
    0,01 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 090,00 €
    3 090,00 €
    1 * 1 ST
       3 090,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    EBRO1340-6612
    Model
    EBI 12-T241
    Sensor
    PT1000, class A
    Radial, 1 external temperature probe, bendeble, Ø 1,5 mm, 500 mm
    Protection class
    IP 68
    Recording intervals
    250 ms to 24 hrs.
    Weight
    90 g
    Data memory - No. of readings
    100,320
    Batteries
    Lithium, 2× BR1225A, 3 V user–replaceable
Software and readout station
Product Information
  • Catalog #
    Description
    Availability
    Price
  • SI 1200, 4-port Interface IF 200, Software Winlog.pro
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 580,00 €
    2 580,00 €
    1 * 1 ST
       2 580,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • SI 1100, Interface IF 100, software Winlog.pro
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 060,00 €
    2 060,00 €
    1 * 1 ST
       2 060,00 €
Complementary Products
Product Information
  • Catalog #
    Accuracy
    Description
    Range
    Resolution
    Availability
    Price
  • EBRO1340-6610
    EBI 12-T222 Temperature logger, 1-channel, radial, 40 mm rigid metal probe
    34
    Log in for availability
    1 * 1 ST - 1 640,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Battery replacement set EBI 12; 3× battery, 6× O-ring, 1× unscrewing aid EBI 12
    Battery replacement set EBI 12; 3× battery, 6× O-ring, 1× unscrewing aid EBI 12
    EBRO1100-0130
    Log in for availability
    1 * 1 ST - 251,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Battery replacement set EBI 12; 3× battery, 6× O-ring
    Battery replacement set EBI 12; 3× battery, 6× O-ring
    620-1693
    Log in for availability
    1 * 1 ST - 176,00 €
Product Information
  • Catalog #
    Availability
    Price
  • AL 1102, Inlay for data logger sets made of plastic for EBI 16 and EBI 12 data logger and interface
    AL 1102, Inlay for data logger sets made of plastic for EBI 16 and EBI 12 data logger and interface
    EBRO1248-1102
    Log in for availability
    1 * 1 ST - 176,00 €
Product Information
  • Catalog #
    Availability
    Price
  • AL 128, Carrying case systainer 1,25 HU
    AL 128, Carrying case systainer 1,25 HU
    EBRO1248-0128
    Log in for availability
    1 * 1 ST - 180,00 €
Product Information
  • Catalog #
    Availability
    Price
  • AL 1100, Inlay for data logger sets made of plastic for EBI 12 data logger and interface
    AL 1100, Inlay for data logger sets made of plastic for EBI 12 data logger and interface
    EBRO1248-1100
    Log in for availability
    1 * 1 ST - 176,00 €
Product Information
  • Catalog #
    Availability
    Price
  • EBI NI - 140, Grommets for fixing sensors
    EBI NI - 140, Grommets for fixing sensors
    EBRO1340-1988
    Log in for availability
    1 * 100 ST - 251,00 €
Product Information
  • Catalog #
    Availability
    Price
  • AL 1103, Inlay for data logger sets made of plastic for EBI 12 data logger and interface
    AL 1103, Inlay for data logger sets made of plastic for EBI 12 data logger and interface
    EBRO1248-1103
    Log in for availability
    1 * 1 ST - 176,00 €
Product Information
  • Catalog #
    Availability
    Price
  • AL 102, Silicone protection for EBI 12 radial, 2 channels
    AL 102, Silicone protection for EBI 12 radial, 2 channels
    EBRO1340-6022
    Log in for availability
    1 * 1 ST - 237,00 €
Product Information
  • Catalog #
    Availability
    Price
  • AL 190, Silicone cable strap set consisting of 10 pcs
    AL 190, Silicone cable strap set consisting of 10 pcs
    EBRO1248-0190
    Log in for availability
    1 * 1 ST - 49,70 €
Product Information
  • Catalog #
    Availability
    Price
  • AL 1101, Inlay for data logger sets made of plastic for EBI 12 data logger
    AL 1101, Inlay for data logger sets made of plastic for EBI 12 data logger
    EBRO1248-1101
    Log in for availability
    1 * 1 ST - 176,00 €
Product Information
  • Catalog #
    Availability
    Price
  • AL 121, Opening tool for EBI 12
    AL 121, Opening tool for EBI 12
    EBRO1100-0131
    Log in for availability
    1 * 1 ST - 48,20 €
Product Information
  • Catalog #
    Availability
    Price
  • EBI-KV3, Compression fitting
    EBI-KV3, Compression fitting
    628-9104
    Log in for availability
    1 * 1 ST - 131,00 €
Product Information
  • Catalog #
    Availability
    Price
  • AL 122, Eyelet for EBI 12 Data logger consisting of 1 eyelid and 2 sealing O-rings
    AL 122, Eyelet for EBI 12 Data logger consisting of 1 eyelid and 2 sealing O-rings
    EBRO1248-0122
    Log in for availability
    1 * 1 ST - 48,80 €