Order Entry
Austria
ContactUsLinkComponent
Magnifier, Linen Tester
Magnifier, Linen Tester
Catalog # 630-8321
Supplier:  ESCHENBACH OPTIK
Magnifier, Linen Tester
Catalog # 630-8321
Supplier:  ESCHENBACH OPTIK

Specifications

  • Ø:
    16 mm
  • Description:
    Optional accessory for linen testers, 8×, 30 ft.
  • Pk:
    1
  • W×D:
    20×20 mm

Specifications

About this item

Thread counter with side scale, measuring thread counter made of plastic. Available for the metal and plastic thread counters.

  • Division of the scale in millimeters
  • Plano-convex PXM® lightweight lens, DUPLEX coated for sharp-edged, distortion-free images
  • Black plastic housing