Order Entry
Austria
ContactUsLinkComponent
 
Thermo Scientific™ Basic Vortex Mixer
Catalog # BARN88882012
undefined
Thermo Scientific™ Basic Vortex Mixer
Catalog # BARN88882012

Specifications

  • Speed range:
    200 - 3000 rpm
  • Description:
    Basic vortex mixer
  • Electrical:
    6 W
  • Speed accuracy:
    ±5% rpm
  • Weight:
    6,5 kg
  • W×D×H:
    205×151×131 mm
  • Pk:
    1

Specifications

About this item

The Basic Vortex Mixer provides comprehensive mixing capabilities for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilution preparation, precipitated assays, cell suspension vortexing and tissue sample mixing.

  • Digital Speed display

  • Continuous and Touch modes
  • Convenient Speed adjustments by knob control
  • Stable for long-term operations
  • Multiple accessories for different vessels