Synthetic Complete Amino Acid Supplement Minus Arginine, Lysine without Yeast Nitrogen Base
USBID9537-17
New Product
:
- Pk:100 g
- Storage temperature:RT/4 °C
- Shipping temperature:RT
Drop-out mixes are used as a supplement to Drop-out base for making synthetic, complete media formulations in accordance with methods in yeast genetics. All lots are quality control tested using the appropriate yeast strains and transformants.
- Dust-Free: Specially milled powder blends are designed to eliminate or greatly reduce airborne particulates
- Homogeneous: Proprietary blending technology ensures complete mixing of components
- Appearance: White, homogeneous, free flowing powder
- Solubility: Colourless, clear, complete
- pH: As Reported