Orders LinkContactUsLinkComponent
KingFisher™ Presto Purification System
KingFisher™ Presto Purification System
KingFisher™ Presto with 96 DW head, 96 samples per run, 5...115 °C heating tempertarure
Catalog # FINN5400830
KingFisher™ Presto Purification System
Catalog # FINN5400830
KingFisher™ Presto Purification System
KingFisher™ Presto with 96 DW head, 96 samples per run, 5...115 °C heating tempertarure
Price
56 500,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories.
  • Volume range of 50 to 5000 μl
  • Minimised size for easy fitting to automation systems
  • Heating capability for application requiring elevated temperatures
  • Easy connection to several liquid handling instruments
  • Flexible system for wide variety of applications
Utilising proven KingFisher magnetic particle-based technology, the KingFisher Presto provides high-quality yields of target nucleic acids and proteins for biopharma, biotech and research.
Specifications
  • Description:
    KingFisher™ Presto with 96 DW head, 96 samples per run, 5...115 °C heating tempertarure
  • Cat. No.:
    FINN5400830
  • Processing volume:
    50 to 1000 μl
  • W×D×H:
    360×465×400 mm
  • Plate types:
    96, 96 DW
  • Weight:
    24 kg