PHAGO'GEL Disinfectant Gel in Airless Bottle Pump
PHAGSA0135720
New Product
- Capacity:1000 ml
- Description:PHAGO'GEL SPS Disinfectant gel with Airless Bottle Pump, Clear
- Sterility:Sterile
- Pk:12
Intended for the hygienic surgical disinfection of the hands by friction.
- High tolerance
- Speed of use
- Bactericidal, fungicidal, mycobactericidal and virucidal
- Fragrance-free, dye-free, phenoxyethanol-free, and paraben-free
It is used as an alternative to simple or hygienic hand washing, in the absence of a water point, during serial contact, before any aseptic gesture.