Order Entry
Austria
ContactUsLinkComponent
Measuring Cylinders, Tall Form, Class A
Measuring Cylinders, Tall Form, Class A
Catalog # WITG3.715.000
Supplier:  witeg Labortechnik
New Product
Measuring Cylinders, Tall Form, Class A
Catalog # WITG3.715.000
Supplier:  witeg Labortechnik
New Product

Specifications

  • NS:
    10/19
  • Capacity:
    5 ml
  • Pk:
    10
  • Tolerance:
    ±0,05 ml

Specifications

About this item

Brown Graduated cylinder class A with spout tall form made of borosilicate glass 3.3/DURAN® components with PE stopper.

Certifications: According to DIN ISO 4788