ContactUsLinkComponent
EasySep™ Human Extracellular Vesicle (CD9) Positive Selection Kit
EasySep™ Human Extracellular Vesicle (CD9) Positive Selection Kit
Catalog # STMC17894
undefined
EasySep™ Human Extracellular Vesicle (CD9) Positive Selection Kit
Catalog # STMC17894
Specifications
  • Description:
    EasySep™ human extracellular vesicle (CD9) positive selection kit, 20 ml biofluid processed
  • Pk:
    1 KIT
Specifications
About this item
Fast and easy isolation of human extracellular vesicles using immunomagnetic positive selection.
  • Fast, easy to use, and column-free
  • Avoid the need for ultracentrifugation and associated time consuming EV isolation methods
  • Obtain highly purified EVs without contaminating biofluid components
The EasySep™ Human Extracellular Vesicle (CD9) Positive Selection Kit isolates highly purified human extracellular vesicles from plasma/serum and cell culture conditioned medium by immunomagnetic positive selection. Extracellular vesicles are labelled with Tetrameric Antibody Complexes recognising CD9 and magnetic particles. labelled extracellular vesicles are separated without columns using an EasySep™ magnet. Unwanted biofluid components are simply poured off, while desired extracellular vesicles remain in the tube. Isolated extracellular vesicles are available for downstream applications such as DNA/RNA extraction, Western blot, or mass spectrometry.​
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').