EasySep™ Human Cord Blood CD34 Positive Selection Kit III
Supplier: STEMCELL Technologies
Certificates
About this item
Complete kit for CD34+ cells from cord blood.
- Fast and easy-to-use
- Up to 96% purity
- No columns required
- Can be combined with SepMate™ for consistent, high-throughput sample processing
Isolate highly purified CD34+ cells from fresh whole umbilical cord blood is recommended.
If isolating CD34+ cells from fresh blood or buffy coat, the Complete Kit for Human Whole Blood CD34+ Cells is recommended. If isolating CD34+ cells from other samples, including fresh or previously frozen mobilised peripheral blood or bone marrow mononuclear cells, or from previously frozen cord blood mononuclear cells, we recommend using the EasySep™ Human CD34 Positive Selection Kit II.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Frequently Bought Together