ContactUsLinkComponent
Pierce™ Blocking Buffers, Protein Free
Pierce™ Blocking Buffers, Protein Free
Catalog # PIER37570
CAS Number:  
undefined
Pierce™ Blocking Buffers, Protein Free
Catalog # PIER37570
CAS Number:  
Specifications
  • Pk:
    1 L
  • Description:
    Pierce™ protein- free (TBS) blocking buffer
  • Seq:
    04
Specifications
About this item
These protein-free blocking buffers are either a PBS or a TBS formulation of a non protein compound that provides effective blocking for membrane based and plate based protein detection methods, resulting in extremely low background.
  • Streptavidin friendly that is absolutely free of biotin
  • Effective for all kinds of protein detection systems
  • Optimized and validated in many protein methods to provide high signal-to-noise ratio
  • Protein free blocker that minimizes or eliminates cross reactivity associated with protein-based blocking buffers
These buffers are devoid of protein while remaining highly effective at blocking plates, membranes and other surfaces for ELISA, western blotting, glass slide arrays and other applications.
Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store product at room temperature. After opening, store product at 4˚C.