Orders LinkContactUsLinkComponent
EasySep™ Mouse T Cell Isolation Kits
EasySep™ Mouse T Cell Isolation Kits
Marketsource
EasySep™ Mouse T Cell Isolation Kits
Marketsource
Certificates
EasySep™ Mouse T Cell Isolation Kits
EasySep™ Mouse T Cell Isolation Kits
2 Options Available Below
Product Details & Documents
Immunomagnetic isolation of untouched mouse T cells
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-T cells and streptavidin-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
This product replaces the EasySep™ Mouse T Cell Enrichment Kit (Catalog #19751) for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Product Family Options
Product Information
  • Availability
    Price
  • STMC19851
    EasySep™ mouse T cell isolation kit, 1×10⁹ cells
    EasySep™ mouse T cell isolation kit, 1×10⁹ cells
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 1 040,00 €
    1 040,00 €
    1 * 1 KIT
       1 040,00 €
    Specifications:
    Marketsource
    Marketsource
  • STMC19851RF
    EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips, 1×10⁹ cells
    EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips, 1×10⁹ cells
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 1 130,00 €
    1 130,00 €
    1 * 1 KIT
       1 130,00 €
    Specifications:
    Marketsource
    Marketsource