ContactUsLinkComponent
EasySep™ Mouse MDSC (CD11b+Gr1+) Isolation Kit
EasySep™ Mouse MDSC (CD11b+Gr1+) Isolation Kit
Catalog # STMC19867
undefined
EasySep™ Mouse MDSC (CD11b+Gr1+) Isolation Kit
Catalog # STMC19867
Specifications
  • Description:
    EasySep™ mouse MDSC (CD11b+Gr1+) isolation kit, 1×10⁹ cells
  • Pk:
    1
Specifications
About this item
Immunomagnetic isolation of untouched myeloid-derived suppressor cells (CD11b+Gr1+).
  • Fast, easy-to-use and column-free
  • Up to 96% purity
  • Untouched, viable cells
The EasySep™ Mouse MDSC (CD11b+Gr1+) Isolation Kit is designed to isolate CD11b+Gr1+ myeloid-derived suppressor cells (MDSCs) from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-MDSCs and streptavidin-coated magnetic particles (RapidSpheres™). labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, cell sorting, and cell culture.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').