ContactUsLinkComponent
RoboSep™-C Tubing Set
RoboSep™-C Tubing Set
Catalog # STMC200-0090
undefined
RoboSep™-C Tubing Set
Catalog # STMC200-0090
Specifications
  • Description:
    RoboSep™-C tubing set, 1 unit
  • Pk:
    1
Specifications
About this item
For automated cell isolation with RoboSep™-C
The RoboSep™-C Tubing Set is a single-use biocompatible tubing set designed for use with RoboSep™-C (Catalog #100-0186), the fully automated closed cell isolation system. The easy-to-install tubing set provides a closed, sterilised fluid path that enables efficient cell washing and cell isolation.
For information on how to install the RoboSep™-C Tubing Set, refer to the RoboSep™-C User Reference Manual or contact us.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').