ContactUsLinkComponent
RoboSep™-C Human T Cell Isolation Kit
RoboSep™-C Human T Cell Isolation Kit
Catalog # STMC100-0204
undefined
RoboSep™-C Human T Cell Isolation Kit
Catalog # STMC100-0204
Specifications
  • Description:
    RoboSep™-C human T cell Isolation kit, 1×10¹⁰ cells
  • Pk:
    1 KIT
Specifications
About this item
Immunomagnetic negative isolation of untouched T cells from leukopaks.
  • Fully automated large-scale isolation in a closed system
  • Up to 99% purity with high recovery
  • Untouched, viable cells
The RoboSep™-C Human T Cell Isolation Kit is designed to isolate untouched and highly purified T cells from fresh human peripheral blood leukopaks using RoboSep™-C (STMC100-0185 and #100-0186) for automated immunomagnetic negative selection.
This kit targets non-T cells for removal with antibodies recognising specific cell surface markers. Using RoboSep™-C in a fully automated protocol, unwanted cells are labeled with antibodies and magnetic particles, and separated without columns using a RoboSep™-C magnet. The desired untouched cells are washed and concentrated by RoboSep™-C and transferred to a final product bag. Isolated cells are immediately available for downstream applications such as flow cytometry, culture, or cryopreservation.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).