ContactUsLinkComponent
EasySep™ Extracellular Vesicle PE Positive Selection Kit
EasySep™ Extracellular Vesicle PE Positive Selection Kit
Catalog # STMC100-0812
undefined
EasySep™ Extracellular Vesicle PE Positive Selection Kit
Catalog # STMC100-0812
Specifications
  • Description:
    EasySep™ extracellular vesicle PE positive selection kit, 1 kit
  • Pk:
    1 KIT
Specifications
About this item
Fast and easy isolation of extracellular vesicles using immunomagnetic positive selection.
  • Fast and easy-to-use
  • No columns required
Efficiently obtain specific extracellular vesicle (EV) subtypes for use in downstream analyses or to enhance the sensitivity of your biomarker discovery assays. Use EasySep™ Extracellular Vesicle PE Positive Selection Kit to isolate tissue- and cell-specific EV subtypes from a variety of biofluids or cell-culture-conditioned medium in just 70 minutes.
EasySep™ Extracellular Vesicle PE Positive Selection Kit can be used with your choice of PE-conjugated antibodies to immunomagnetically isolate EV subtypes based on EV-associated surface markers. Compared to bulk EV isolation techniques (e.g. differential ultracentrifugation), the fast, easy, and column-free protocol results in higher enrichment of EV subtypes, making it easier to detect low frequency EVs in downstream analyses. Isolated EVs remain intact and are compatible with downstream analyses such as DNA/RNA extraction, western blot, or mass spectrometry.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).