ContactUsLinkComponent
RosetteSep™ Human ILC2 Enrichment Kit
RosetteSep™ Human ILC2 Enrichment Kit
  STMC15382
undefined
RosetteSep™ Human ILC2 Enrichment Kit
  STMC15382
  • Description:
    RosetteSep™ human ILC2 enrichment kit, 200 ml whole blood processed
  • Pk:
    1 KIT
Immunodensity enrichment of untouched human ILC2s.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
  • Facilitates rapid flow sorting of ILC2s
The RosetteSep™ Human ILC2 Enrichment Cocktail is designed to enrich human ILC2s from whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-ILC2s and glycophorin A on red blood cells (RBCs). When centrifuged over a density gradient medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The ILC2 cells are present as a highly enriched population at the interface between the plasma and the density gradient medium.
 : Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).