Immunomagnetic positive selection of human CD19+ cells (e.g. B cells).
- Fast and easy-to-use
- Up to 99% purity
- No columns required
The EasySep™ Human CD19 Positive Selection Kit II is designed to isolate CD19+ cells from fresh or previously frozen peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimise nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is expressed on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.
This product replaces the EasySep™ Human CD19 Positive Selection Kit (STMC18054) for even faster cell isolations.