
Potassium permanganate 0.02M (0.1N), meets analytical specification of Ph.Eur. standard volumetric solution
Catalog # 474370010.
Supplier: Thermo Fisher Scientific
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Appearance:purple-black liquid
- Strength:0.0998 to 0.1002 N
Specification Test Results
About this item
:
:158.03 g/mol
Merck Index:13,00774
Density:1
EINECS:231-760-3
:GHS07
Storage Temperature:Ambient
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364