Order Entry
Austria
ContactUsLinkComponent
2-Propanol solution in water 70%, technical
Flammable
Irritant
2-Propanol solution in water 70%, technical
Certificates
Flammable
Irritant
2-Propanol solution in water 70%, technical
Certificates

Specification Test Results

  • Appearance:
    clear colorless liquid
  • Concentration:
    69 to 71 % (determined via difference in water content by KF)
  • Water:
    29 to 31 %
  • Density:
    0.858 to 0.862 kg/l

Specification Test Results

Related Information

Frequently Bought Together