
Doxapram hydrochloride hydrate 98+%
New Product
:7081-53-0
:
:C₂₄H₃₀N₂O₂·HCl·H₂O
:MFCD09954698
:432.98 g/mol
:13,03465
:Ambient
:2811
:6.1,III
:GHS06
- Appearance:white to almost white crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥98 %
- Loss on drying:3 % to 4.5 % (1 g, 105°C, 2 h)
- Titration with NaOH:98.0 to 102.0 % (on dry substance)
Frequently Bought Together