
(Z)-4-Hydroxytamoxifen 99%
Catalog # 469501000.
Supplier: Thermo Fisher Scientific
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:white to almost white crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
- Melting point:137°C to 143°C
Specification Test Results
About this item
:
:387.51 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:68047-06-3
Formula:C₂₆H₂₉NO₂
MDL Number:MFCD00941389