QPix® XE Microbial Colony Pickers
About this item
The QPix® XE is a microbial colony picker ideal for labs with reduced space, but need efficient colony picking throughput. The QPix® XE is a great entry-level system to replace highly subjective and error prone manual picking with automated colony picking.
- Easy-to-use software
- High-picking efficiency
- Screen and pick colonies
- Automatic tracking
The QPix® microbial colony pickers combine intelligent image analysis with precise automation for fast and efficient screening of large libraries. With a variety of data tracking and assay tools, the QPix® Software streamlines the control and management of complex and iterative processes.
The QPix® XE is ideal for small lab spaces with no compromise on efficiency, with advanced imaging and robotic capabilities. The modular platform allows for further integration into an automated work cell for throughput growth or walk-away time. The QPix® XE has a picking capacity of 1500 colonies per hr white light and 1000 colonies per hour in fluorescence mode.
Delivery information: Includes dedicated PC with pre-loaded control software, standard picking head, 1 year warranty covering parts and labor.
2 Options Available Below
Product Details & Documents
The QPix® XE is a microbial colony picker ideal for labs with reduced space, but need efficient colony picking throughput. The QPix® XE is a great entry-level system to replace highly subjective and error prone manual picking with automated colony picking.
- Easy-to-use software
- High-picking efficiency
- Screen and pick colonies
- Automatic tracking
The QPix® microbial colony pickers combine intelligent image analysis with precise automation for fast and efficient screening of large libraries. With a variety of data tracking and assay tools, the QPix® Software streamlines the control and management of complex and iterative processes.
The QPix® XE is ideal for small lab spaces with no compromise on efficiency, with advanced imaging and robotic capabilities. The modular platform allows for further integration into an automated work cell for throughput growth or walk-away time. The QPix® XE has a picking capacity of 1500 colonies per hr white light and 1000 colonies per hour in fluorescence mode.
Delivery information: Includes dedicated PC with pre-loaded control software, standard picking head, 1 year warranty covering parts and labor.
Documents
Product Family Options
Product Information
- AvailabilityPrice
- MLDVX9500/220-AWESQPix® XE colony picking system (220 V) - white light colony picker
- MLDVX9500F/220AWESQPix® XE fluorescent colony picking system (220 V) - white light and fluorescence colony pickerQPix® XE fluorescent colony picking system (220 V) - white light and fluorescence colony pickerLog in for availability1 * 1 ST