Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Bottle Closures
Screw Caps, DIN 45, Black
Screw Caps, DIN 45, Black
Screw Caps, DIN 45, Black
Catalog # RIXIU8S1001451400
Supplier:
RIXIUS
New Product
Expand image
Screw Caps, DIN 45, Black
Catalog # RIXIU8S1001451400
Supplier:
RIXIUS
New Product
Specifications
Thread:
GL 45
Colour:
Black
Description:
Screw cap, sealing of closure LDPE
Material:
PP
Pk:
1.400
Specifications
About this item
Product Details
Product Details