Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonia - Ammonium Chloride Buffer Solution, pH 10

Ammonia - Ammonium Chloride Buffer Solution, pH 10

Ammonia - ammonium chloride buffer solution, 10,0, 1 L
Catalog # ACSDCH8004008.1L
Supplier:   CPACHEM
Ammonia - Ammonium Chloride Buffer Solution, pH 10
Catalog # ACSDCH8004008.1L
Supplier:  CPACHEM
CAS Number:  
Ammonia - Ammonium Chloride Buffer Solution, pH 10

Ammonia - ammonium chloride buffer solution, 10,0, 1 L

CAS Number:  
Price
107,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Ammonia - Ammonium chloride buffer solution.

  • Shelf life: 12 months
  • Storage temperature: 15 to 25 °C

Certifications: ISO 9001

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Ammonia - ammonium chloride buffer solution
  • pH value:
    10,0

Specifications