Order Entry
Austria
ContactUsLinkComponent
Sulphuric acid diluted
Sulphuric acid diluted
Catalog # ACSDCH8002119.1L
Supplier:  CPACHEM
New Product
CAS Number:  
Sulphuric acid diluted
Catalog # ACSDCH8002119.1L
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :98.08 g/mol
Storage Temperature:Room Temperature
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589