Order Entry
Austria
ContactUsLinkComponent
Tin, AAS Standard Solution
Tin, AAS Standard Solution
Catalog # ACSDA055.20CP.L1
Supplier:  CPACHEM
New Product
CAS Number:  
Tin, AAS Standard Solution
Catalog # ACSDA055.20CP.L1
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    20% HCl
  • Pk:
    100 mL
  • Storage conditions:
    15...25 °C
  • Shelf life:
    36 Months
  • Element:
    Tin
  • Symbol:
    Sn

Specifications

About this item

Certifications: ISO 17034, ISO 17025, ISO 9001