Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M7 Printers
Catalog # BRDYM7-17-498
Supplier: Brady
New Product
Specifications
- Colour:White
- Dimensions:25,4×12,7 mm
- Material:Vinyl
- Model:M7-17-498
- Packed:500
- Pk:1 Roll
- Adhesive:Repositionable, rubber based
Specifications
About this item
Permanent or repositionable, once applied the B-498 CleanLift® Series Vinyl Cloth remains in place, until it must be changed.
- Adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning
- Sticks to any clean, dry surface
- Specially formulated topcoat for excellent print quality
- Semi-gloss finish for crisp, clean look
Labels are ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6200.