Cannabicyclolic Acid (CRM)
CAYM35745-1
New Product
:
- Pk:1 mg
- CAS No.:40524-99-0
- Description:Cannabicyclolic acid (CRM)
- Shipping conditions:Wet ice in continental US; may vary elsewhere
- Synonym(s):CBLA
- Formula:C₂₂H₃₀O₄
- Molecular weight:358,5
- Storage temperature:–20 °C
(±)-Cannabicyclolic acid (CRM) is a certified reference material categorized as a photodegradation product of the phytocannabinoid cannabichromenic acid.
: This product is intended for research and forensic applications.